Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt Chloride Hexahydrate Cell Culture MP Biomedicals

Catalog No. ICN19464225 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19464225 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19464225 Supplier MP Biomedicals, Inc Supplier No. 0219464225
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Reagent-grade chemical intended for research and further manufacturing applications.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7791-13-1
Molecular Formula Cl2CoH12O6
Molecular Weight (g/mol) 237.92
MDL Number MFCD00149652
InChI Key GFHNAMRJFCEERV-UHFFFAOYSA-L
Synonym Cobalt Dichloride Hexahydrate, Cobaltous Chloride, Hexahydrate
IUPAC Name λ²-cobalt(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Co++]

Specifications

Quantity 25 g
Density 1.920 g/mL
Formula Weight 237.9
Grade Cell Culture
Chemical Name or Material Cobalt Chloride, Hexahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.