Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt Chloride Hexahydrate, MP Biomedicals™

Catalog No. MP219403590 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219403590 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP219403590 Supplier MP Biomedicals, Inc Supplier No. 0219403590
Add to Cart
Add to Cart

Cobalt chloride is an inorganic compound of cobalt and chlorine.

  • ACS Grade
  • Cobalt chloride an excellent indicator for water in dessicants
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7791-13-1
Molecular Formula Cl2CoH12O6
Molecular Weight (g/mol) 237.92
MDL Number MFCD00149652
InChI Key GFHNAMRJFCEERV-UHFFFAOYSA-L
Synonym Cobalt (2+) Chloride Hexahydrate, Cobaltous Chloride Hexahydrate, Chlorek cobaltawy, Cobalt(II) chloride hexahydrate
IUPAC Name λ²-cobalt(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Co++]

Specifications

Physical Form Crystalline Powder
Quantity 500 g
Assay Percent Range 98.0–102.0%
Solubility Information Soluble in water.
Formula Weight 237.93g/mol
Percent Purity 99.6%
Grade ACS
Chemical Name or Material Cobalt Chloride Hexahydrate
Recommended Storage Store at Room Temperature (15–30°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.