Learn More
Perkin Elmer US LLC Cobalt (Co) Internal Standard - Sulfur-Free, 3%, 400 g
Catalog No. 502104527
Encompass_Preferred
Matrix: Concentration: Volume: Content: Hydrocarbon Oil 3% 400 g Cobalt (Sulfur-Free)
Catalog No. 50-210-4527 Supplier Perkin Elmer US LLC Supplier No. N9304168
Add to Cart
Add to Cart
Request a Quote
Matrix: Concentration: Volume: Content: Hydrocarbon Oil 3% 400 g Cobalt (Sulfur-Free)