Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt(II) acetate, anhydrous, 98+%

Catalog No. aab2321818
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2321818 50 g
AAB2321830 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2321818 Supplier Thermo Scientific Chemicals Supplier No. B2321818
Only null left
Add to Cart

CAS: 71-48-7 | C4H6CoO4 | 177.02 g/mol

Cobalt(II) acetate is used as an industrial catalyst. It is used as a precursor to various oil drying agents. It finds application in ion exchange agents, lubricants, plating agents and surface treating agents, greases, ink, toner, and colorant products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cobalt(II) acetate is used as an industrial catalyst. It is used as a precursor to various oil drying agents. It finds application in ion exchange agents, lubricants, plating agents and surface treating agents, greases, ink, toner, and colorant products.

Solubility
Soluble in water, alcohol and dilute acids.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-48-7
Molecular Formula C4H6CoO4
Molecular Weight (g/mol) 177.02
MDL Number MFCD00008689
InChI Key QAHREYKOYSIQPH-UHFFFAOYSA-L
Synonym cobalt ii acetate, cobalt diacetate, cobalt acetate, cobaltous diacetate, bis acetato cobalt, cobalt 2+ acetate, cobaltous acetate, acetic acid, cobalt 2+ salt, cobalt di acetate, cobalt 2+ diacetate
PubChem CID 6277
ChEBI CHEBI:85138
IUPAC Name cobalt(2+);diacetate
SMILES [Co++].CC([O-])=O.CC([O-])=O

Specifications

Melting Point 298°C (decomposition)
Odor Odorless
Linear Formula Co(OOCCH3)2
Quantity 50 g
UN Number UN3077
Sensitivity Hygroscopic
Merck Index 14,2433
Solubility Information Soluble in water,alcohol and dilute acids.
Formula Weight 177.02
Percent Purity ≥98%
Chemical Name or Material Cobalt(II) acetate, anhydrous
Show More Show Less
Hazard Category H317-H334-H341-H350i-H360F
Hazard Statement GHS H Statement
H334-H350-H360-H341-H302-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Suspected of causing genetic defects.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 200-755-8
RTECSNumber AG3150000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.