Learn More
Strem, An Ascensus Company CAS# 6147-53-1. 100g. Cobalt(II) acetate tetrahydrate, 98+%. MFCD00149569
SDP
Catalog No. 5090114706
Encompass
CAS# 6147-53-1. 100g. Cobalt(II) acetate tetrahydrate, 98+%. MFCD00149569. Molecular Weight: 177.00 (249.08). Molecular Formula: Co(OOCCH3)2∙4H2O. Color/form: pink xtl. Strem# 93-2701. http://www.strem.com/catalog/v/93-2701/
Catalog No. 50-901-14706 Supplier Strem, An Ascensus Company Supplier No. 932701100G
CAS# 6147-53-1. 100g. Cobalt(II) acetate tetrahydrate, 98+%. MFCD00149569. Molecular Weight: 177.00 (249.08). Molecular Formula: Co(OOCCH3)2∙4H2O. Color/form: pink xtl. Strem# 93-2701. http://www.strem.com/catalog/v/93-2701/