Learn More
Sigma Aldrich Fine Chemicals Biosciences Cobalt(II) acetate tetrahydrate reagent grade | Purity: 23.0-25.0% Co basis (EDTA titration) | Mol Wt: 249.08 | 6147-53-1 | MFCD00149569 | 50G
Catalog No. 501879419
Encompass
Cobalt(II) acetate tetrahydrate reagent grade | Purity: 23.0-25.0% Co basis (EDTA titration) | Mol Wt: 249.08 | 6147-53-1 | MFCD00149569 | 50G
Catalog No. 50-187-9419 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 20839650G
Cobalt(II) acetate tetrahydrate reagent grade | Purity: 23.0-25.0% Co basis (EDTA titration) | Mol Wt: 249.08 | 6147-53-1 | MFCD00149569 | 50G