Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt(II) acetate tetrahydrate, 98%

Catalog No. AAA1837422
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1837422 100 g
AAA1837414 25 g
AAA1837436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1837422 Supplier Thermo Scientific Chemicals Supplier No. A1837422
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 6147-53-1 | C4H14CoO8 | 249.081 g/mol

Cobalt(II) acetate tetrahydrate is used as an industrial catalyst to harden paints and varnishes, an active catalyst for oxidation and esterification reactions. It is also used to prepare metal complexes having unusual coordination geometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cobalt(II) acetate tetrahydrate is used as an industrial catalyst to harden paints and varnishes, an active catalyst for oxidation and esterification reactions. It is also used to prepare metal complexes having unusual coordination geometry.

Solubility
Soluble in water, alcohol, dilute acids and phenyl acetate.

Notes
Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6147-53-1
Molecular Formula C4H14CoO8
MDL Number MFCD00149569
InChI Key ZBYYWKJVSFHYJL-UHFFFAOYSA-L
Synonym cobalt ii acetate tetrahydrate, cobalt acetate tetrahydrate, cobaltous acetate tetrahydrate, cobalt diacetate tetrahydrate, bis acetato tetraquacobalt, octan kobaltnaty czech, acetic acid, cobalt 2+ salt, tetrahydrate, octan kobaltnaty, ccris 9441
PubChem CID 61372

Specifications

Melting Point ∼140°C (in -4H2O)
Density 1.71
Odor Odorless
Refractive Index 1.542
Linear Formula Co(OOCCH3)2·4H2O
Quantity 100 g
UN Number UN3077
Beilstein 3731397
Sensitivity Hygroscopic
Merck Index 14,2433
Solubility Information Soluble in water,alcohol,dilute acids and phenyl acetate.
SMILES CC(=O)[O-].CC(=O)[O-].O.O.O.O.[Co+2]
IUPAC Name cobalt(2+);diacetate;tetrahydrate
Molecular Weight (g/mol) 249.081
Formula Weight 249.08 (177.02 Anhydrous)
Percent Purity 98%
Chemical Name or Material Cobalt(II) acetate tetrahydrate
Show More Show Less
Hazard Category H302-H317-H334-H335-H341-H350i-H360F
Hazard Statement GHS H Statement
H334-H350-H360-H341-H302-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Suspected of causing genetic defects.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P271-P272-P280g-P281-P285-P302+P352-P304+P340-P308+P313-P333+P313-P342+P311-P363-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 200-755-8
RTECSNumber AG3325000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.