Alfa Aesar™ Cobalt(II) acetate tetrahydrate, ACS, 98.0-102.0%

Manufacturer: Alfa Aesar™ 4434414

Quantity 25g

 View more versions of this product

Catalog No. AA4434414

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6147-53-1
    Molecular Formula C4H6CoO4·4H2O
    Linear Formula Co(OOCCH3)2·4H2O
    Formula Weight 249.08 (177.02 Anhydrous)
    MDL Number MFCD00149569
    UN Number UN3077
    Beilstein 3731397
    Merck Index 14,2433
    Chemical Name or Material Cobalt (II) acetate tetrahydrate
    Grade ACS Grade

    Specifications

    Formula Weight 249.08 (177.02 Anhydrous)
    Melting Point ≈140°C (in -4H2O)
    Health Hazard 2 GHS H Statement
    H334-H350-H360-H341-H302-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause cancer.
    May damage fertility or the unborn child.
    Suspected of causing genetic defects.
    Harmful if swallowed.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P284a-P261-P280-P342+P311-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    CAS 6147-53-1
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    Refractive Index 1.542
    Molecular Formula C4H6CoO4·4H2O
    Linear Formula Co(OOCCH3)2·4H2O
    Quantity 25g
    Sensitivity Hygroscopic
    MDL Number MFCD00149569
    Merck Index 14,2433
    Color Red
    RTECS Number AG3325000
    Odor Odorless
    UN Number UN3077
    Physical Form Crystalline
    Beilstein 3731397
    Density 1.71g/mL
    Chemical Name or Material Cobalt (II) acetate tetrahydrate
    Assay 98.0 to 102.0%
    Grade ACS Grade

    GHS H Statement
    H334-H350-H360-H341-H302-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause cancer.
    May damage fertility or the unborn child.
    Suspected of causing genetic defects.
    Harmful if swallowed.
    May cause an allergic skin reaction.

    GHS P Statement
    P284a-P261-P280-P342+P311-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit