Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt(II) bromide, ultra dry, 99.99% (metals basis)

Catalog No. AA4031903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4031903 1 g
AA4031906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4031903 Supplier Thermo Scientific Chemicals Supplier No. 04031903
Add to Cart

CAS: 7789-43-7 | Br2Co | 218.741 g/mol

Used primarily as a catalyst in some processes. Chiefly used in hygrometers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-43-7
Molecular Formula Br2Co
Molecular Weight (g/mol) 218.741
MDL Number MFCD00016017
InChI Key BZRRQSJJPUGBAA-UHFFFAOYSA-L
Synonym cobalt bromide, cobalt ii bromide, cobalt bromide cobr2, cobalt dibromide, cobaltous bromide, hsdb 998, cobalt bromide ous
PubChem CID 24610
IUPAC Name dibromocobalt
SMILES [Co](Br)Br

Specifications

Melting Point 678°C (in N2)
Color Green
Physical Form Beads
Quantity 1 g
Assay Percent Range 99.99% (Metal basis)
Solubility Information Soluble in water. Soluble in methyl acetate,ether,alcohol,acetone.
Formula Weight 218.75
Sensitivity Hygroscopic
Appearance Green (powder/flake/crystalline/beads, etc.)
Density 4.909 g/cm3
Chemical Name or Material Cobalt(II) bromide
Show More Show Less
Hazard Category H302-H315-H317-H319-H334-H350
Precautionary Statement P201-P202-P261-P264b-P270-P272-P280-P281-P285-P301+P312-P302+P352-P304+P341-P305+P351+P338-P308+P313-P330-P333+P313-P342+P311-P362-P501c
EINECSNumber 232-166-7
RTECSNumber GF9595000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.