Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt(II) chloride, anhydrous, 99.7% (metals basis)

Catalog No. AA1230318
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1230318 50 g
AA1230330 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1230318 Supplier Thermo Scientific Chemicals Supplier No. 01230318
Add to Cart

Sensitivity: Hygroscopic; 3.367. Decomposes at 400°C on long heating in air | CAS: 7646-79-9 | Cl2Co | 129.83 g/mol

Cobalt(II) chloride is used in humidity indicator in weather instruments. In the anhydrous form, it finds use in electroplating of cobalt, in organic chemistry and is a precursor to cobaltocene, (bis(cyclopentadienyl)cobalt(II), which is a good reducing agent. It also serves as a Lewis acid. Cobalt chloride is an indicator for water in desiccants, owing to the reversible hydration/dehydration coupled with distinct color change. Cobalt chloride is useful for producing invisible ink as it turns blue when heated and becomes invisible once it gets cooled. Cobalt(II) chloride catalyzes cross coupling of aryl halides or vinyl halides with aryl grignard reagents in excellent yields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7646-79-9
Molecular Formula Cl2Co
Molecular Weight (g/mol) 129.83
MDL Number MFCD00010938
InChI Key GVPFVAHMJGGAJG-UHFFFAOYSA-L
Synonym cobalt ii chloride, cobaltchloride, cobalt chloride 0.1 m solution, cobalt ii chloride, anhydrous, cobalt ii-chloride, curator_000013, cobalt chloride anhydrous, wln: co g2, cobalt-ii-chloride
IUPAC Name λ²-cobalt(2+) dichloride
SMILES [Cl-].[Cl-].[Co++]

Specifications

Boiling Point 1049°C
Melting Point 735°C
Color Blue
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.7% (Metals basis)
Solubility Information Freely soluble in water and methanol. Soluble in alcohol,acetone,ether,glycerol,and pyridine.
Formula Weight 129.84
Odor Odorless
Sensitivity Hygroscopic
Density 3.367
Chemical Name or Material Cobalt(II) chloride, Anhydrous
Show More Show Less
Hazard Category H302+H332-H317-H318-H334-H335-H341-H350i-H360F-H373
Hazard Statement Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P330-P333+P313-P342+P311-P363-P501c
DOTInformation DOT Class: 9, Packaging Group: III
EINECSNumber 231-589-4
RTECSNumber GF9800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.