Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cobalt(II) chloride hexahydrate

Catalog No. 769495100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-949-5100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 76-949-5100G Supplier Sigma Aldrich Supplier No. 769495100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Cobalt (II) chloride finds applications in electroplating and catalyst preparation. It is also used as precursor for synthesis of electrode materials for lithium ion batteries.

Specifications

CAS 7791-13-1
Molecular Formula Cl2Co -+ 6H2O
Linear Formula CoCl2 -+ 6H2O
MDL Number MFCD00149652
Quantity 100 g
Synonym Cobaltous chloride hexahydrate
Molecular Weight (g/mol) 237.93
Percent Purity ≥97%; 97.0-102.0% (KT)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.