Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt(II) chloride hexahydrate, ACS, 98.0-102.0%

Catalog No. p-4407399
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3655422 100 g
AA3655436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3655422 Supplier Thermo Scientific Chemicals Supplier No. 03655422
Only null left
Add to Cart

CAS: 7791-13-1 | Cl2CoH12O6 | 237.92 g/mol

Cobalt chloride is used as a humidity indicator in weather instruments. It finds use in electroplating of cobalt, in invisible ink, in organic chemistry and is a precursor to cobaltocene(bis (cyclopentadienyl) cobalt(II)).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7791-13-1
Molecular Formula Cl2CoH12O6
Molecular Weight (g/mol) 237.92
MDL Number MFCD00149652
InChI Key GFHNAMRJFCEERV-UHFFFAOYSA-L
IUPAC Name λ²-cobalt(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Co++]

Specifications

Melting Point 87°C
Color Purple-Red
Physical Form Crystalline
Quantity 100 g
Assay Percent Range 98.0-102.0%
Solubility Information Soluble in water,alcohol,acetone,ether and glycerol.
Formula Weight 237.93 (129.84 Anhydrous)
Grade ACS
Odor Odorless
Sensitivity Hygroscopic
Appearance Red-purple crystals
Density 1.924 g/cm3
Chemical Name or Material Cobalt(II) chloride hexahydrate, For analysis ACS
Show More Show Less
Hazard Category H302+H332-H317-H334-H341-H350i-H360F
Precautionary Statement P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P340-P308+P313-P330-P333+P313-P342+P311-P363-P501c
DOTInformation DOT Class: 9, Packaging Group: III
EINECSNumber 231-589-4
RTECSNumber GG0200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.