Learn More
Cobalt(II) chloride hexahydrate, for analysis
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 86.0°C |
| Color | Purple to Red |
| Physical Form | Powder |
| Quantity | 1 kg |
| Linear Formula | CoCl2·6H2O |
| Fieser | 01,155; 06,127; 11,479 |
| Merck Index | 15, 2421 |
| Solubility Information | Solubility in water: 970g/L (20°C) |
| Formula Weight | 237.93 |
| Percent Purity | 98.0 to 102.0% (Complexometry) |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement May damage fertility. Harmful if inhaled. Harmful if swallowed. Suspected of causing genetic defects. May cause cancer by inhalation. May cause an allergic skin reaction. May cause allergy or ast |
| Precautionary Statement | GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF ON SKIN: |
| RTECSNumber | GG0200000 |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.