Learn More
Strem, An Ascensus Company CAS# [866-81-9]. 250g. Cobalt(II) citrate hydrate. MFCD00150521
SDP
Supplier:  Strem, An Ascensus Company 932707250G
Encompass
CAS# [866-81-9]. 250g. Cobalt(II) citrate hydrate. MFCD00150521. Molecular Weight: 555.01. Molecular Formula: Co3(C6H5O7)2∙XH2O. Color/form: light purple pwdr. Strem# 93-2707. www.strem.com/catalog/v3/93-2707/
Catalog No. 50-197-9393