Learn More
Cobalt(II) fluoride, anhydrous, 98%
Description
Cobalt(II) fluoride can be used as a catalyst to alloy metals. It is also used for optical deposition, of which it tremendously improves optical quality. The compound may be used in dental care.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 1400°C |
| Melting Point | 1200°C |
| Physical Form | Powder |
| Quantity | 100 g |
| UN Number | UN2923 |
| Merck Index | 14,2440 |
| Solubility Information | Soluble in water and hydrofluoric acid. insoluble in alcohol,ether,benzene. |
| Formula Weight | 96.93 |
| Percent Purity | 98% |
| Odor | Odorless |
| Show More |
Safety and Handling
| Hazard Category | H301-H314 |
| Hazard Statement | GHS H Statement H301-H314-H351-H317 Toxic if swallowed. Causes severe skin burns and eye damage. Suspected of causing cancer. May cause an allergic skin reaction. |
| Precautionary Statement | P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
| DOTInformation | Hazard Class: 8; Packaging Group: II |
| EINECSNumber | 233-061-9 |
| RTECSNumber | GG0770000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.