Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt(II) fluoride, ultra dry, 99.99% (metals basis)

Numéro de catalogue. AA3574009
Modifier l'affichage
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AA3574009 10 g
AA3574004 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AA3574009 Fournisseur Thermo Scientific Chemicals Code fournisseur 03574009
Ajouter au panier
Ajouter au panier
Request Bulk or Custom Formats

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

CAS: 10026-17-2 | CoF2 | 96.93 g/mol

It is used as a catalyst to alloy metals and also used for optical deposition, of which it tremendously improves optical quality, in dental care.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 10026-17-2
Molecular Formula CoF2
Molecular Weight (g/mol) 96.93
MDL Number MFCD00010941
InChI Key YCYBZKSMUPTWEE-UHFFFAOYSA-L
Synonym cobalt ii fluoride, cobalt fluoride, cobalt difluoride, cobaltous fluoride, hsdb 236, cobalt fluoride pink, cobalt ii fluoride, powder, cobalt ii fluoride, anhydrous, cobalt ii fluoride, ultra dry
PubChem CID 24820
IUPAC Name difluorocobalt
SMILES F[Co]F

Spécifications

Boiling Point 1400°C
Melting Point 1200°C
Physical Form Powder
Quantity 10 g
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic
Density 4.46 g/mL
UN Number UN2923
Merck Index 14,2440
Solubility Information Soluble in water,alcohol and acetone.
Formula Weight 96.93
Percent Purity 99.99%
Assay (metals basis)
Chemical Name or Material Cobalt(II) fluoride, Ultra dry
Afficher plus de résultats Afficher moins de résultats
Hazard Category H301-H314
Hazard Statement GHS H Statement
H301-H314-H351-H317
Toxic if swallowed.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause an allergic skin reaction.
Precautionary Statement P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 233-061-9
RTECSNumber GG0770000
TSCA Yes
missing translation for 'storageNote'
  • Ampouled under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only

missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'

missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.