Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt(II) hexafluoro-2,4-pentanedionate hydrate, 97%, Thermo Scientific™

Catalog No. AA4159403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4159403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4159403 Supplier Thermo Scientific Chemicals Supplier No. 4159403

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 206986-92-7
Molecular Formula C10H6CoF12O5
Molecular Weight (g/mol) 493.07
MDL Number MFCD00150649,MFCD00042510
InChI Key POHWVOSIFCILEF-UHFFFAOYSA-N
Synonym bis hexafluoroacetylacetonato cobalt ii hydrate, cobalt 2+ bis 1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate, cobalt 2+ ; z-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate
PubChem CID 16212172
IUPAC Name cobalt;(Z)-1,1,1,5,5,5-hexafluoro-4-hydroxypent-3-en-2-one;hydrate
SMILES O.[Co].OC(=CC(=O)C(F)(F)F)C(F)(F)F.OC(=CC(=O)C(F)(F)F)C(F)(F)F

Specifications

Melting Point 173°C to 179°C
Color Orange
Physical Form Powder
Quantity 1 g
Solubility Information It is insoluble in water.
Formula Weight 473.04 (Anhydrous)
Percent Purity 97%
Odor Odorless
Chemical Name or Material Cobalt(II) hexafluoro-2,4-pentanedionate hydrate
Hazard Category H315-H317-H319-H335-H350
Hazard Statement GHS H Statement
H351-H315-H319-H317-H335
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P271-P272-P280-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P333+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.