Learn More
Strem, An Ascensus Company CAS# 21041-93-0. 50g. Cobalt(II) hydroxide (97%-Co). MFCD00016015
SDP
Catalog No. 5090114744
Encompass
CAS# 21041-93-0. 50g. Cobalt(II) hydroxide (97%-Co). MFCD00016015. Molecular Weight: 92.96. Molecular Formula: Co(OH)2. Color/form: pink pwdr. Strem# 93-2741. http://www.strem.com/catalog/v/93-2741/
Catalog No. 50-901-14744 Supplier Strem, An Ascensus Company Supplier No. 93274150G
CAS# 21041-93-0. 50g. Cobalt(II) hydroxide (97%-Co). MFCD00016015. Molecular Weight: 92.96. Molecular Formula: Co(OH)2. Color/form: pink pwdr. Strem# 93-2741. http://www.strem.com/catalog/v/93-2741/