Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt(II,III) oxide, 99% (metals basis), Thermo Scientific™

Catalog No. AA4466014
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4466014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4466014 Supplier Thermo Scientific Chemicals Supplier No. 4466014

missing translation for 'validMainframeMsg'

S.A. 40-70m2/g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Color Black
CAS 1308-06-1
Physical Form Powder
Quantity 25 g
Assay Percent Range 99% (Metal basis)
Molecular Formula Co3O4
MDL Number MFCD00010939
Synonym oxocobalt; oxo oxocobaltiooxy cobalt, cobalt ii oxide; cobalt iii oxide, cobalt ii, coo.co2o3, aronis24129, cobaltic tetraoxide,, cobalt ii,iii oxide, powder, <10 mum, cobalt ii oxide; oxo oxocobaltio oxy cobalt, cobalt ii,iii oxide metals basis , 2-6 micron powder, oxidanylidenecobalt; oxidanylidene oxidanylidenecobaltiooxy cobalt
InChI Key UBEWDCMIDFGDOO-UHFFFAOYSA-N
SMILES [O--].[O--].[O--].[O--].[Co++].[Co+3].[Co+3]
IUPAC Name dicobalt(3+) λ²-cobalt(2+) tetraoxidandiide
Molecular Weight (g/mol) 240.80
PubChem CID 11651651
Formula Weight 240.8
Odor Odorless
Appearance Black powder
Density 6.11g/cm3
Chemical Name or Material Cobalt(II, III) oxide
Show More Show Less
Hazard Category H351-H400-H410-H317; Suspected of causing cancer. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. May cause an allergic skin reaction.
DOTInformation DOT Class: 9, Packaging Group: III
EINECSNumber 215-157-2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.