Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt(II) iodide, ultra dry, 99.999% (metals basis)

Catalog No. AA3573006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3573006 5 g
AA3573003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3573006 Supplier Thermo Scientific Chemicals Supplier No. 03573006
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 15238-00-3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15238-00-3
MDL Number MFCD00016030
Synonym cobalt ii iodide, acmc-20ale6, cobalt ii iodide, anhydrous, cobalt ii iodide, ultra dry, cobalt ii iodide, powder, cobalt ii iodide, anhydrous, powder trace metals basis

Specifications

Boiling Point 570°C (vac.)
Melting Point 515°C (vac.)
Color White
Physical Form Beads
Quantity 5 g
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic; Light sensitive
Density 5.584 g/mL
Merck Index 14,2443
Formula Weight 312.74
Percent Purity 99.999%
Assay (metals basis)
Chemical Name or Material Cobalt(II) iodide, Ultra dry
Show More Show Less
Hazard Category H317-H341-H350
Hazard Statement GHS H Statement
H341-H317
Suspected of causing genetic defects.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
EINECSNumber 239-283-2
TSCA Yes
storageNote1
  • Ampouled under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.