Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt(II) iodide, ultra dry, 99.999% (metals basis)

Catalog No. AA4718503
Change view
Click to view available options
Quantity:
1 g
Packaging:
Ampouled under argon
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AA4718503 1 g Ampouled under argon
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4718503 Supplier Thermo Scientific Chemicals Supplier No. 04718503
Add to Cart

CAS: 15238-00-3 | CoI2

Cobalt(II) iodide is used as a moisture and humidity indicator. It is also a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15238-00-3
Molecular Formula CoI2
MDL Number MFCD00016030
Synonym cobalt ii iodide, acmc-20ale6, cobalt ii iodide, anhydrous, cobalt ii iodide, ultra dry, cobalt ii iodide, powder, cobalt ii iodide, anhydrous, powder trace metals basis

Specifications

Melting Point 515°C (vac.)
Boiling Point 570°C (vac.)
Packaging Ampouled under argon
Quantity 1 g
Sensitivity Hygroscopic; Light sensitive
Solubility Information Soluble in water.
Formula Weight 312.74
Percent Purity 99.999%
Physical Form Powder
Chemical Name or Material Cobalt(II) iodide
Hazard Category H317-H341-H350
Precautionary Statement P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
EINECSNumber 239-283-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.