Learn More
Strem, An Ascensus Company CAS# 10026-22-9. 500g. Cobalt(II) nitrate hexahydrate, 99%. MFCD00149647
SDP
Catalog No. 5090114730
Encompass
CAS# 10026-22-9. 500g. Cobalt(II) nitrate hexahydrate, 99%. MFCD00149647. Molecular Weight: 182.99 (291.05). Molecular Formula: Co(NO3)2∙6H2O. Color/form: red xtl. Strem# 93-2730. http://www.strem.com/catalog/v/93-2730/
Catalog No. 50-901-14730 Supplier Strem, An Ascensus Company Supplier No. 932730500G
CAS# 10026-22-9. 500g. Cobalt(II) nitrate hexahydrate, 99%. MFCD00149647. Molecular Weight: 182.99 (291.05). Molecular Formula: Co(NO3)2∙6H2O. Color/form: red xtl. Strem# 93-2730. http://www.strem.com/catalog/v/93-2730/