Learn More
Strem, An Ascensus Company CAS# 10026-22-9. 25g. Cobalt(II) nitrate hexahydrate (99.999%-Co) PURATREM. MFCD00149647
SDP
Catalog No. 5090114751
Encompass
CAS# 10026-22-9. 25g. Cobalt(II) nitrate hexahydrate (99.999%-Co) PURATREM. MFCD00149647. Molecular Weight: 182.99 (291.05). Molecular Formula: Co(NO3)2∙6H2O. Color/form: red to purple xtl. Strem# 93-2746. http://www.strem.com/catalog/v/93-2746/
Catalog No. 50-901-14751 Supplier Strem, An Ascensus Company Supplier No. 93274625G
CAS# 10026-22-9. 25g. Cobalt(II) nitrate hexahydrate (99.999%-Co) PURATREM. MFCD00149647. Molecular Weight: 182.99 (291.05). Molecular Formula: Co(NO3)2∙6H2O. Color/form: red to purple xtl. Strem# 93-2746. http://www.strem.com/catalog/v/93-2746/