Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cobalt(II) nitrate hexahydrate

Catalog No. 20310610G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-310-610G 10 g
20-310-650G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 20-310-610G Supplier Sigma Aldrich Supplier No. 20310610G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Cobalt(II) nitrate hexahydrate

Specifications

CAS 10026-22-9
Melting Point 55°C
Molecular Formula CoN2O6 -+ 6H2O
Linear Formula Co(NO3)2 -+ 6H2O
Quantity 10 g
MDL Number MFCD00149647
Synonym Cobaltous nitrate; Cobaltous nitrate hexahydrate; Nitric acid, cobalt(II) salt
Molecular Weight (g/mol) 291.03
Percent Purity 99.999% trace metals basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.