Cobalt(II) oxide, 95%, Alfa Aesar™

Manufacturer: Alfa Aesar 2314122

Quantity 100g

 View more versions of this product

Catalog No. AA2314122

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1307-96-6
    Physical Form Powder
    Assay Percent Range 95%
    Formula Weight 74.93
    MDL Number MFCD00016031
    Chemical Name or Material Cobalt(II) oxide

    Specifications

    Formula Weight 74.93
    Quantity 100g
    EINECS Number 215-154-6
    Physical Form Powder
    Density 6.45
    Assay Percent Range 95%
    MDL Number MFCD00016031
    CAS 1307-96-6
    Chemical Name or Material Cobalt(II) oxide

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit