Learn More
Cobalt(II) oxide, 99.995% (metals basis)
Description
Cobalt(II) oxide is widely used in the ceramic industry. It is an additive to create blue-colored glazes and enamels. It finds application in the manufacture of magnetic and electronic component materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 1935°C |
| Color | Brown-Green |
| Physical Form | Powder |
| Quantity | 100 g |
| UN Number | UN2811 |
| Merck Index | 14,2446 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 74.93 |
| Percent Purity | 99.995% |
| Concentration or Composition (by Analyte or Components) | CoO, may contain up to 10% Co3O4 |
| Show More |
Safety and Handling
| Hazard Category | H301-H317-H330-H334-H350 |
| Hazard Statement | GHS H Statement H351-H302-H317 Suspected of causing cancer. Harmful if swallowed. May cause an allergic skin reaction. |
| Precautionary Statement | P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P284-P285-P301+P310-P302+P352-P304+P340-P308+P313-P310-P330-P333+P313-P363-P501c |
| DOTInformation | Hazard Class: 6.1; Packaging Group: II |
| EINECSNumber | 215-154-6 |
| RTECSNumber | GG2800000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.