Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cobalt(II) phthalocyanine

Catalog No. 30769650G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-769-650G 50 g
30-769-610G 10 g
30-769-61G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 30-769-650G Supplier Sigma Aldrich Supplier No. 30769650G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Cobalt(II) phthalocyanine (CoPC )is a metallopthalocyanine.Self assembled molecules of CoPC were deposited on epitaxial graphene. CoPC may be used to develop carbon black based electrocatalysts. CoPC modified carbon paste may be used as indicators for electrocatalytic amperometric measurements. ZnO impregnated with CoPC may be used as a sensitizer to determine the photodegradation of cyanide in aqueous suspension. CoPC catalysed aerobic regenerations of aldehydes and ketones have been investigated. CoPC may also be used as an end capping agent of a hyperbranched poly(aryl ether ketone) to be used for oxidative decomposition of 2,4,6,-trichlorophenol.

Specifications

CAS 3317-67-7
Molecular Formula C32H16CoN8
Linear Formula C32H16CoN8
Quantity 50 g
MDL Number MFCD00010718
Synonym Cobaltous phthalocyaninate; PcCo; Phthalocyanine cobalt(II) salt
Molecular Weight (g/mol) 571.46
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.