Learn More
Strem, An Ascensus Company CAS# 10026-24-1. 100g. Cobalt(II) sulfate heptahydrate, 98+%. MFCD00149658
SDP
Catalog No. 5090114752
Encompass
CAS# 10026-24-1. 100g. Cobalt(II) sulfate heptahydrate, 98+%. MFCD00149658. Molecular Weight: 155.00 (281.10). Molecular Formula: CoSO4∙7H2O. Color/form: pink to red xtl. Strem# 93-2749. http://www.strem.com/catalog/v/93-2749/
Catalog No. 50-901-14752 Supplier Strem, An Ascensus Company Supplier No. 932749100G
CAS# 10026-24-1. 100g. Cobalt(II) sulfate heptahydrate, 98+%. MFCD00149658. Molecular Weight: 155.00 (281.10). Molecular Formula: CoSO4∙7H2O. Color/form: pink to red xtl. Strem# 93-2749. http://www.strem.com/catalog/v/93-2749/