Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt(Ii) Sulfate Heptahydrate, Solstice

Catalog No. 6000427 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-004-27 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-004-27 Supplier Solstice Supplier No. 12933500G

missing translation for 'validMainframeMsg'

Purum,≥95%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Specifications

CAS 10026-24-1
Quantity 500 g
Molecular Formula CoH14O11S
Linear Formula CoSO4 · 7H2O
MDL Number MFCD00149658
UN Number 3077
Synonym cobalt sulfate heptahydrate, cobalt 2+ sulfate heptahydrate, cobaltous sulfate heptahydrate, cobalt ii sulfate heptahydrate, cobalt monosulfate heptahydrate, dsstox_cid_340, dsstox_rid_75523, dsstox_gsid_20340, bieberite, coo4s.7h2o
InChI Key MEYVLGVRTYSQHI-UHFFFAOYSA-L
SMILES O.O.O.O.O.O.O.[Co++].[O-]S([O-])(=O)=O
IUPAC Name cobalt(2+);sulfate;heptahydrate
Molecular Weight (g/mol) 281.09
PubChem CID 61444
Formula Weight 281.1g/mol
Percent Purity ≥95%
Grade purum
Density 2.03g/mL at 25°C
Chemical Name or Material Cobalt(Ii) Sulfate Heptahydrate
Show More Show Less
Hazard Statement P201-P261-P280-P284-P304 + P340-P308 + P313
Precautionary Statement H302-H317-H334-H341-H350i-H360F-H410
EINECSNumber 233-334-2
RTECSNumber GG3200000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.