Learn More
Strem, An Ascensus Company CAS# 1317-42-6. 25g. Cobalt(II) sulfide (99.5%-Co). MFCD00016032
SDP
Catalog No. 5090114757
Encompass
CAS# 1317-42-6. 25g. Cobalt(II) sulfide (99.5%-Co). MFCD00016032. Molecular Weight: 91.00. Molecular Formula: CoS. Color/form: -200 mesh black pwdr. Strem# 93-2751. http://www.strem.com/catalog/v/93-2751/
Catalog No. 50-901-14757 Supplier Strem, An Ascensus Company Supplier No. 93275125G
CAS# 1317-42-6. 25g. Cobalt(II) sulfide (99.5%-Co). MFCD00016032. Molecular Weight: 91.00. Molecular Formula: CoS. Color/form: -200 mesh black pwdr. Strem# 93-2751. http://www.strem.com/catalog/v/93-2751/