Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cobalt(II) TPP | 14172-90-8 | ≥98.0% | 671.65 g/mol | C44H28CoN4 | 1 G
SDP

Catalog No. 5000210239
Encompass_Preferred

Cobalt(II) TPP (5,10,15,20-tetraphenylporphyrin cobalt(II)) is a metalloporphyrin reagent used in research applications including catalysis, sensing, and materials chemistry. It is supplied as a purple solid with high purity and is suitable for laboratory and analytical use.

  • High purity (≥98.0%) suitable for research use.
  • Molecular weight 671.65 g/mol and formula C44H28CoN4.
  • Commonly used in catalysis, oxygen reduction, and sensing studies.
  • Soluble in common organic solvents; purple solid.
  • Available as 1 G unit for laboratory experiments.

Catalog No. 50-002-10239 Supplier Medchemexpress LLC Supplier No. HYW0730131G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cobalt(II) TPP (5,10,15,20-tetraphenylporphyrin cobalt(II)) is a metalloporphyrin reagent used in research applications including catalysis, sensing, and materials chemistry. It is supplied as a purple solid with high purity and is suitable for laboratory and analytical use.

  • High purity (≥98.0%) suitable for research use.
  • Molecular weight 671.65 g/mol and formula C44H28CoN4.
  • Commonly used in catalysis, oxygen reduction, and sensing studies.
  • Soluble in common organic solvents; purple solid.
  • Available as 1 G unit for laboratory experiments.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.