Learn More
Strem, An Ascensus Company CAS# 21679-46-9. 10g. Cobalt(III) acetylacetonate, 98+%. MFCD00013488
SDP
Catalog No. 5090114709
Encompass
CAS# 21679-46-9. 10g. Cobalt(III) acetylacetonate, 98+%. MFCD00013488. Molecular Weight: 356.30. Molecular Formula: Co(CH3COCHCOCH3)3. Color/form: green xtl. Strem# 93-2702. http://www.strem.com/catalog/v/93-2702/
Catalog No. 50-901-14709 Supplier Strem, An Ascensus Company Supplier No. 93270210G
CAS# 21679-46-9. 10g. Cobalt(III) acetylacetonate, 98+%. MFCD00013488. Molecular Weight: 356.30. Molecular Formula: Co(CH3COCHCOCH3)3. Color/form: green xtl. Strem# 93-2702. http://www.strem.com/catalog/v/93-2702/