Learn More
Description
Specifications
Specifications
| CAS | 61789-51-3 |
| Quantity | 250 g |
| MDL Number | MFCD02094763 |
| Biological Activity | Central nervous system, Gastrointestinal tract |
| Refractive Index | n20/D 1.4620 |
| Density | 0.921 g/mL (at 25°C) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.