Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt Naphthenate (Co ca. 10%), TCI America™
SDP

Catalog No. N033825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N033825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N033825G Supplier TCI America Supplier No. N033825G

Chemical Identifiers

CAS 61789-51-3
Molecular Formula C20H34CoO4
Molecular Weight (g/mol) 397.42
MDL Number MFCD02094763
InChI Key XSWKLHINRKWMTD-UHFFFAOYNA-L
Synonym cobalt naphthenate, cobalt ii 2-naphthoate
PubChem CID 119077305
IUPAC Name λ²-cobalt(2+) bis(3-(3-ethylcyclopentyl)propanoate)
SMILES [Co++].CCC1CCC(CCC([O-])=O)C1.CCC1CCC(CCC([O-])=O)C1

Specifications

Melting Point 140°C
Color Black-Purple
Physical Form Crystalline/Lump
Quantity 25 g
UN Number 2001
Chemical Name or Material Cobalt Naphthenate (Co ca. 10%)
EINECSNumber (8)-0630
RTECSNumber QK8925000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.