Learn More
Electron Microscopy Sciences Cobalt Naphthenate Solution 10 ML
SDP
Catalog No. 50980294
Encompass_Preferred
Solution of 55% Cobalt Napthenate and 45% Mineral Spirit that is used as an activator for the Vestopal embedding resin.
Catalog No. 50-980-294 Supplier Electron Microscopy Sciences Supplier No. 12500
Solution of 55% Cobalt Napthenate and 45% Mineral Spirit that is used as an activator for the Vestopal embedding resin.