Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt Nitrate ACS AR Crystal, Macron™

Catalog No. MK4544500 Shop All Avantor Products
Change view
Click to view available options
Glass; 500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MK-4544-500 Glass; 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK-4544-500 Supplier Avantor Supplier No. 454404

missing translation for 'validMainframeMsg'

Specifications

CAS 10026-22-9
Physical Form Crystalline
Quantity 500 g
Molecular Formula CoH12N2O12
MDL Number MFCD00149647
Synonym cobalt nitrate hexahydrate, cobalt ii nitrate hexahydrate, cobaltous nitrate hexahydrate, cobalt dinitrate hexahydrate, cobalt 2+ nitrate hexahydrate, cobaltous nitrate, hexahydrate, kobalt ii-nitrat-hexahydrat, nitric acid, cobalt 2+ salt, hexahydrate
InChI Key QGUAJWGNOXCYJF-UHFFFAOYSA-N
SMILES O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
IUPAC Name λ²-cobalt(2+) hexahydrate dinitrate
Molecular Weight (g/mol) 291.03
PubChem CID 24821
ChEBI CHEBI:86214
Formula Weight 293.03
Grade ACS
Packaging Glass
Chemical Name or Material Cobalt Nitrate, 6-Hydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.