Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Cobalt Nitrate Solution, 1M
SCI_ED

Catalog No. S25259 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S25259 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25259

missing translation for 'validMainframeMsg'

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 10141-05-6 , 7732-18-5
Molecular Formula CoN2O6
Molecular Weight (g/mol) 182.941
InChI Key UFMZWBIQTDUYBN-UHFFFAOYSA-N
Synonym cobalt nitrate, cobaltous nitrate, cobalt ii nitrate, cobalt dinitrate, cobalt bis nitrate, cobalt 2+ nitrate, nitric acid, cobalt 2+ salt, unii-65w79bfd5v, cobalt nitrate co no3 2, hsdb 238
PubChem CID 25000
ChEBI CHEBI:86209
IUPAC Name cobalt(2+);dinitrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Co+2]

Specifications

Density 1.15g/mL at 20°C
Formula Weight 291.03
Packaging 1M
Quantity 50 mL
Chemical Name or Material Cobalt Nitrate Solution, 1M, 1.0M
Safety handling Check Alert storage code Red
ShelfLife 2 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.