Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Cobalt(ous) Chloride, Hexahydrate
SCI_ED

Catalog No. s25257 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Lab Grade
Reagent Grade
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25257 100 g Lab Grade
S25257A 100 g Reagent Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25257
Only null left
Add to Cart
Edge
Add to Cart

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 7791-13-1 , 7732-18-5
Molecular Formula Cl2CoH12O6
Molecular Weight (g/mol) 237.92
MDL Number MFCD00149652
InChI Key GFHNAMRJFCEERV-UHFFFAOYSA-L
Synonym Cobalt(II) Chloride Hexahydrate, Cobalt muriate hexahydrate
IUPAC Name λ²-cobalt(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Co++]

Specifications

Color Purple
Physical Form Solid
Quantity 100 g
Assay Percent Range <75%w/v
Linear Formula CoCl2 · 6H2O
Formula Weight 237.95
Grade Reagent
Packaging Lab Grade
Density 1.924
Chemical Name or Material Cobalt(ous) Chloride, Hexahydrate
Safety handling Check Alert storage code Red
ShelfLife 2 years
DOTInformation DOT Class 8, III, : Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.