Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Cobalt(ous) Chloride, Hexahydrate
SCI_ED

Catalog No. S25260 Shop All Fisherbrand Products
Change view
Click to view available options
Concentration or Composition (by Analyte or Components):
1M
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration or Composition (by Analyte or Components)
S25260 1M
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25260

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 7791-13-1 , 7732-18-5
Molecular Formula Cl2CoH12O6
Molecular Weight (g/mol) 237.92
MDL Number MFCD00149652
InChI Key GFHNAMRJFCEERV-UHFFFAOYSA-L
Synonym Cobalt (II) Chloride, Cobalt Muriate Hexahydrate
IUPAC Name λ²-cobalt(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Co++]

Specifications

Color Pink-Purple
Quantity 500 mL
Assay Percent Range <75%w/v
Linear Formula CoCl2·6H2O
Formula Weight 237.95
Concentration or Composition (by Analyte or Components) 1M
Chemical Name or Material Colbalt Chloride Solution
Safety handling Check Alert storage code Red
ShelfLife 2 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.