Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt Standard (Xylene, Plasma Grade)
SDP

Catalog No. ORGCOX2Z Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ORG-COX-2Z 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ORG-COX-2Z Supplier SPEX CertiPrep Supplier No. ORGCOX2Z

Chemical Identifiers

CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Packaging Poly Bottle
Certificate of Analysis With Certificate of Analysis.
Quantity 50 g
Grade Plasma/Xylene
Concentration or Composition (by Analyte or Components) Matrix Concentration Cat. No.
Xylene 1000PPM ORG-COX-2Z
Xylene 5000PPM ORG-COX-4Z
Chemical Name or Material Cobalt Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.