Learn More
Hach Company Cobalt Standard Solution, 1000 mg/L as Co (NIST), 100 mL
Catalog No. 501995475 Shop All Hach Company Products
Encompass
For accuracy checks in cobalt determinations. (HNO3 matrix)
Catalog No. 50-199-5475 Supplier Hach Company Supplier No. 2150342
For accuracy checks in cobalt determinations. (HNO3 matrix)