Learn More
Strem, An Ascensus Company CAS# 14172-90-8. 250MG. Cobalt(II) meso-tetraphenylporphine. MFCD00010725
SDP
Catalog No. 501963561
Encompass
CAS# 14172-90-8. 250MG. Cobalt(II) meso-tetraphenylporphine. MFCD00010725. Molecular Formula: (C44H28N4)Co. Molecular Weight: 671.66. Color/Form: rust colored xtl. Strem# 27-0470. www.strem.com/catalog/v3/27-0470/
Catalog No. 50-196-3561 Supplier Strem, An Ascensus Company Supplier No. 270470250MG
CAS# 14172-90-8. 250MG. Cobalt(II) meso-tetraphenylporphine. MFCD00010725. Molecular Formula: (C44H28N4)Co. Molecular Weight: 671.66. Color/Form: rust colored xtl. Strem# 27-0470. www.strem.com/catalog/v3/27-0470/