Learn More
Cayman Chemical Cobaltic(III) Protoporphyrin IX Chloride, Purity: ≥95%, CAS: 102601-60-5, 100 mg
SDP
Catalog No. 502947374
Encompass_Preferred
An inducer of HO-1 activity; induces HO-1 activity in vitro and in vivo; reduces δ-aminolevulinate synthase and CYP activities in rat liver microsomes ex vivo at 125 µmol/kg; prevents liver injury and increases in the percentage of hepatic apoptotic cells in a mouse model of liver ischemia-reperfusion injury at 5 mg/kg; increases survival and decreases the number of lung CFUs in mice infected with Y. pestis; has also been used as a catalyst for the electrochemical reduction of carbon dioxide in carbon electrodes
Catalog No. 50-294-7374 Supplier Cayman Chemical Supplier No. 33794100 MG
An inducer of HO-1 activity; induces HO-1 activity in vitro and in vivo; reduces δ-aminolevulinate synthase and CYP activities in rat liver microsomes ex vivo at 125 µmol/kg; prevents liver injury and increases in the percentage of hepatic apoptotic cells in a mouse model of liver ischemia-reperfusion injury at 5 mg/kg; increases survival and decreases the number of lung CFUs in mice infected with Y. pestis; has also been used as a catalyst for the electrochemical reduction of carbon dioxide in carbon electrodes