Learn More
Cayman Chemical CobaltIc III ProtprhyrIn 10mg
SDP
Catalog No. 502948506
Encompass_Preferred
An inducer of HO-1 activity; induces HO-1 activity in vitro and in vivo; reduces δ-aminolevulinate synthase and CYP activities in rat liver microsomes ex vivo at 125 µmol/kg; prevents liver injury and increases in the percentage of hepatic apoptotic cells in a mouse model of liver ischemia-reperfusion injury at 5 mg/kg; increases survival and decreases the number of lung CFUs in mice infected with Y. pestis; has also been used as a catalyst for the electrochemical reduction of carbon dioxide in carbon electrodes
Catalog No. 50-294-8506 Supplier Cayman Chemical Supplier No. 3379410 MG
An inducer of HO-1 activity; induces HO-1 activity in vitro and in vivo; reduces δ-aminolevulinate synthase and CYP activities in rat liver microsomes ex vivo at 125 µmol/kg; prevents liver injury and increases in the percentage of hepatic apoptotic cells in a mouse model of liver ischemia-reperfusion injury at 5 mg/kg; increases survival and decreases the number of lung CFUs in mice infected with Y. pestis; has also been used as a catalyst for the electrochemical reduction of carbon dioxide in carbon electrodes