Learn More
Strem, An Ascensus Company CAS# 12427-42-8. 1g. Cobalticinium hexafluorophosphate, 98%. MFCD00011874
SDP
Supplier:  Strem, An Ascensus Company 2704101G
Encompass
CAS# 12427-42-8. 1g. Cobalticinium hexafluorophosphate, 98%. MFCD00011874. Molecular Weight: 334.09. Molecular Formula: (C5H5)2Co+PF6ae. Color/form: yellow xtl. Strem# 27-0410. http://www.strem.com/catalog/v/27-0410/
Catalog No. 50-901-11162