Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobaltous Acetate Tetrahydrate (Crystalline/Certified), Fisher Chemical

Catalog No. C364500 Shop All Fisherbrand Products
Change view
Click to view available options
Glass Bottle; 500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
C364-500 Glass Bottle; 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. C364-500

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 140°C
Color Red
CAS 6147-53-1
pH 6.8
Physical Form Solid
Quantity 500 g
Molecular Formula C4H14CoO8
Linear Formula Co(CH3COO)2·4H2O
MDL Number MFCD00149569
UN Number 3077
Synonym cobalt ii acetate tetrahydrate, cobalt acetate tetrahydrate, cobaltous acetate tetrahydrate, cobalt diacetate tetrahydrate, bis acetato tetraquacobalt, octan kobaltnaty czech, acetic acid, cobalt 2+ salt, tetrahydrate, octan kobaltnaty, ccris 9441
InChI Key ZBYYWKJVSFHYJL-UHFFFAOYSA-L
SMILES CC(=O)[O-].CC(=O)[O-].O.O.O.O.[Co+2]
IUPAC Name cobalt(2+);diacetate;tetrahydrate
Molecular Weight (g/mol) 249.081
PubChem CID 61372
Formula Weight 249.08
Grade Certified
Identification Pass Test
Packaging Glass Bottle
Density 380g/mL
Chemical Name or Material Cobaltous Acetate Tetrahydrate
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
May cause cancer by inhalation. May impair fertility. May cause skin, eye, and respiratory tract irritation. May cause allergic respiratory and skin reaction. Possible risks of irreversible effects. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Hygroscopic. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention.

NFPA

Health:3
Flammability:1
Instability:1
DOTInformation DOT Class 9, : Miscellaneous
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.