Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobaltous Ammonium Sulfate, MP Biomedicals

Catalog No. ICN211119 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN211119 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN211119 Supplier MP Biomedicals, Inc Supplier No. 0521111980
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity chemical reagent for research or further manufacturing applications.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13596-46-8
Molecular Weight (g/mol) 395.21
Synonym diammonium cobalt ii sulfate hexahydrate, cobalt 2+ diammonium hexahydrate disulfate, cobaltous ammonium sulfate hexahydrate, sulfuric acid, ammoniumcobalt 2+ salt 2:2:1

Specifications

Physical Form Powder
Quantity 100 g
Formula Weight 395.23
Chemical Name or Material Cobaltous Ammonium Sulfate
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.