Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobaltous Chloride CS, USP/EP Red Primary Solution, Ricca Chemical
BIOPHARMASDP

Catalog No. 221016 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
2210-16 500 mL Amber Glass Bottle
2210-4 120 mL Amber Glass Bottle
2210-32 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 2210-16 Supplier Ricca Chemical Company Supplier No. 221016
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

1 mL = 59.5 mg CoCl2.6H2O

Chemical Identifiers

CAS 7791-13-1 , 7647-01-0 , 7732-18-5
Molecular Formula Cl2CoH12O6
Molecular Weight (g/mol) 237.92
MDL Number MFCD00149652
InChI Key GFHNAMRJFCEERV-UHFFFAOYSA-L
IUPAC Name λ2-cobalt(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Co++]

Specifications

Color Red
Equivalence 1 mL = 59.5 mg CoCl2·6H2O
Grade EP/USP/Colorimetric Solution
Packaging Amber Glass Bottle
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Cobaltous Chloride, Red Primary Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.