Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobaltous Chloride TS, 2% (w/v) in dilute Hydrochloric Acid, Ricca Chemical
SDP

Catalog No. 220816 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2208-16 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 2208-16 Supplier Ricca Chemical Company Supplier No. 220816
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7791-13-1 , 7732-18-5 , 7647-01-0
Molecular Formula Cl2CoH12O6
Molecular Weight (g/mol) 237.92
MDL Number MFCD00149652
InChI Key GFHNAMRJFCEERV-UHFFFAOYSA-L
Synonym Cobalt (II) Chloride, Cobalt muriate hexahydrate
PubChem CID 24643
ChEBI CHEBI:53503
IUPAC Name λ2-cobalt(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Co++]

Specifications

Color Pink
CAS Min % 1.9, 95.0, 1.12
CAS Max % 1.98, 99.0, 1.17
Percent Purity 2%
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix Dilute Hydrochloric Acid
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 2% (w/v)
Chemical Name or Material Cobaltous Chloride, Test Solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.