Learn More
Bound Tree Medical Coban™ Self-Adherent Wrap, Tan, 2in W X 5yd L
Catalog No. NC0288817
Encompass_Preferred
Coban self-adherent wrap is a laminate of nonwoven material and elastic fibers placed lengthwise to provide elasticity. The elastic wrap contains a cohesive material that makes it stick to itself but not to other materials or skin. Caution: This Product Contains Natural Rubber Latex Which May Cause Allergic Reactions
Catalog No. NC0288817 Supplier Bound Tree Medical Supplier No. F925607
Coban self-adherent wrap is a laminate of nonwoven material and elastic fibers placed lengthwise to provide elasticity. The elastic wrap contains a cohesive material that makes it stick to itself but not to other materials or skin. Caution: This Product Contains Natural Rubber Latex Which May Cause Allergic Reactions